Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NIT2
Entrez Gene ID
56954
Gene Name
nitrilase family member 2
Gene Aliases
HEL-S-8a
Chromosome Location
Chr.3: 100334718 - 100355634 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020202.4 NP_064587.1 2 - 3 204 136
GenBank mRNA AF260334.1 - 2 - 3 203 136
AF284574.1 - 2 - 3 151 136
AK225979.1 - 2 - 3 180 136
AK294010.1 - 2 - 3 165 136
AK295835.1 - 2 - 3 165 136
AK313704.1 - 2 - 3 146 136
BC020620.1 - 2 - 3 161 136
BC107890.1 - 2 - 3 192 136
DB466042.1 - 2 - 3 204 136
FJ224326.1 - 2 - 3 151 136

View Full Product Details