Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NIT2
Entrez Gene ID
56954
Gene Name
nitrilase family member 2
Gene Aliases
HEL-S-8a
Chromosome Location
Chr.3: 100334718 - 100355634 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020202.4 NP_064587.1 5 - 6 503 73
GenBank mRNA AF260334.1 - 5 - 6 502 73
AF284574.1 - 5 - 6 450 73
AK313704.1 - 5 - 6 445 73
BC020620.1 - 5 - 6 460 73
BC107890.1 - 5 - 6 491 73
FJ224326.1 - 5 - 6 450 73

View Full Product Details