Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FER1L5
Entrez Gene ID
90342
Gene Name
fer-1 like family member 5
Gene Aliases
-
Chromosome Location
Chr.2: 96642719 - 96704887 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293083.1 NP_001280012.1 35 - 36 3994 94
XM_011512110.2 XP_011510412.1 35 - 36 4011 94
XM_011512113.2 XP_011510415.1 35 - 36 4005 94
XM_011512115.2 XP_011510417.1 34 - 35 3942 94
XM_011512116.2 XP_011510418.1 35 - 36 3924 94
XM_011512117.2 XP_011510419.1 34 - 35 3906 94
XM_011512118.2 XP_011510420.1 34 - 35 3891 94
XM_011512119.2 XP_011510421.1 35 - 36 4011 94
XM_011512120.2 XP_011510422.1 34 - 35 3902 94
XM_011512122.2 XP_011510424.1 35 - 36 4012 94
XM_011512124.2 XP_011510426.1 19 - 20 3595 94
XM_011512125.2 XP_011510427.1 35 - 36 4012 94
XM_011512126.2 XP_011510428.1 35 - 36 4011 94
XM_017005222.1 XP_016860711.1 35 - 36 4008 94
XM_017005223.1 XP_016860712.1 18 - 19 2659 94
XM_017005224.1 XP_016860713.1 9 - 10 1451 94
GenBank mRNA AL133022.1 - 17 - 18 3397 94
BC126368.1 - 1 - 2 205 94

View Full Product Details