Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FER1L5
Entrez Gene ID
90342
Gene Name
fer-1 like family member 5
Gene Aliases
-
Chromosome Location
Chr.2: 96642719 - 96704887 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293083.1 NP_001280012.1 41 - 42 4619 58
XM_011512110.2 XP_011510412.1 41 - 42 4636 58
XM_011512111.2 XP_011510413.1 41 - 42 4633 58
XM_011512113.2 XP_011510415.1 41 - 42 4630 58
XM_011512114.2 XP_011510416.1 41 - 42 4633 58
XM_011512115.2 XP_011510417.1 40 - 41 4567 58
XM_011512116.2 XP_011510418.1 41 - 42 4549 58
XM_011512117.2 XP_011510419.1 40 - 41 4531 58
XM_011512118.2 XP_011510420.1 40 - 41 4516 58
XM_011512120.2 XP_011510422.1 40 - 41 4527 58
XM_011512122.2 XP_011510424.1 41 - 42 4637 58
XM_011512124.2 XP_011510426.1 25 - 26 4220 58
XM_017005222.1 XP_016860711.1 41 - 42 4633 58
XM_017005223.1 XP_016860712.1 24 - 25 3284 58
XM_017005224.1 XP_016860713.1 15 - 16 2076 58
GenBank mRNA AL133022.1 - 23 - 24 4022 58
AY461813.1 - 40 - 41 4648 58
BC117324.1 - 7 - 8 827 58
BC126368.1 - 7 - 8 830 58
BC143825.1 - 7 - 8 827 58

View Full Product Details