The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012505.1 | NP_001012523.1 | 5 - 6 | 522 | 117 |
NM_001244808.1 | NP_001231737.1 | 5 - 6 | 625 | 117 | |
NM_001244810.1 | NP_001231739.1 | 5 - 6 | 522 | 117 | |
NM_001244812.1 | NP_001231741.1 | 2 - 3 | 150 | 117 | |
NM_001244816.1 | NP_001231745.1 | 4 - 5 | 427 | 117 | |
NM_032682.5 | NP_116071.2 | 5 - 6 | 522 | 117 | |
XM_005264737.4 | XP_005264794.1 | 5 - 6 | 531 | 117 | |
XM_006713102.2 | XP_006713165.1 | 4 - 5 | 483 | 117 | |
XM_006713103.2 | XP_006713166.1 | 2 - 3 | 220 | 117 | |
XM_006713104.2 | XP_006713167.1 | 3 - 4 | 453 | 117 | |
XM_011533584.2 | XP_011531886.1 | 5 - 6 | 593 | 117 | |
XM_011533585.2 | XP_011531887.1 | 4 - 5 | 445 | 117 | |
XM_017006165.1 | XP_016861654.1 | 4 - 5 | 364 | 117 | |
XM_017006166.1 | XP_016861655.1 | 4 - 5 | 460 | 117 | |
XM_017006168.1 | XP_016861657.1 | 5 - 6 | 531 | 117 | |
GenBank mRNA | AF146696.1 | - | 3 - 4 | 259 | 117 |
AF250920.1 | - | 4 - 5 | 427 | 117 | |
AK122710.1 | - | 5 - 6 | 625 | 117 | |
BC005055.1 | - | 5 - 6 | 472 | 117 | |
BC054815.1 | - | 5 - 6 | 472 | 117 | |
BC071893.1 | - | 5 - 6 | 472 | 117 | |
BC080521.1 | - | 5 - 6 | 472 | 117 | |
BC131720.1 | - | 2 - 3 | 147 | 117 |