Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABHD14B
Entrez Gene ID
84836
Gene Name
abhydrolase domain containing 14B
Gene Aliases
CIB, HEL-S-299
Chromosome Location
Chr.3: 51968510 - 51974630 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC071931.1 - 1 - 2 212 93

View Full Product Details