Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
QARS
Entrez Gene ID
5859
Gene Name
glutaminyl-tRNA synthetase
Gene Aliases
GLNRS, MSCCA, PRO2195
Chromosome Location
Chr.3: 49095932 - 49105129 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272073.1 NP_001259002.1 13 - 14 1528 69
NM_005051.2 NP_005042.1 13 - 14 1561 69
NR_073590.1 13 - 14 1512 69
XM_017006965.1 XP_016862454.1 13 - 14 1241 69
GenBank mRNA AF130067.1 - 9 - 10 689 69
AK056815.1 - 13 - 14 1140 69
AK222510.1 - 13 - 14 1189 69
AK290523.1 - 13 - 14 1189 69
AK293260.1 - 12 - 13 1088 69
AK297987.1 - 12 - 13 1035 69
AK301559.1 - 13 - 14 1157 69
BC000394.2 - 13 - 14 1170 69
BC001567.1 - 13 - 14 1168 69
BC001772.1 - 9 - 10 696 69
BC016634.1 - 12 - 13 993 69
BC029739.1 - 13 - 14 1171 69
X76013.1 - 13 - 14 1170 69

View Full Product Details