Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
QARS
Entrez Gene ID
5859
Gene Name
glutaminyl-tRNA synthetase
Gene Aliases
GLNRS, MSCCA, PRO2195
Chromosome Location
Chr.3: 49095932 - 49105129 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272073.1 NP_001259002.1 21 - 22 2448 78
NM_005051.2 NP_005042.1 21 - 22 2481 78
NR_073590.1 21 - 22 2432 78
XM_017006965.1 XP_016862454.1 21 - 22 2161 78
GenBank mRNA AF130067.1 - 17 - 18 1609 78
AK056815.1 - 21 - 22 2060 78
AK290523.1 - 21 - 22 2109 78
AK293260.1 - 20 - 21 2008 78
AK297987.1 - 20 - 21 1955 78
AK301559.1 - 21 - 22 2077 78
BC000394.2 - 21 - 22 2090 78
BC001567.1 - 21 - 22 2088 78
BC001772.1 - 17 - 18 1616 78
BC016634.1 - 20 - 21 1913 78
BC029739.1 - 21 - 22 2091 78
X76013.1 - 21 - 22 2090 78

View Full Product Details