The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022805.3 | NP_073716.1 | 3 - 4 | 360 | 72 |
NM_022806.3 | NP_073717.1 | 3 - 4 | 370 | 72 | |
NM_022807.3 | NP_073718.1 | 4 - 5 | 505 | 72 | |
NM_022808.3 | NP_073719.1 | 3 - 4 | 397 | 72 | |
GenBank mRNA | AK222633.1 | - | 3 - 4 | 320 | 72 |
DA069028.1 | - | 3 - 4 | 370 | 72 | |
DA131564.1 | - | 3 - 4 | 314 | 72 |