Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HHAT
Entrez Gene ID
55733
Gene Name
hedgehog acyltransferase
Gene Aliases
MART2, SKI1, Skn
Chromosome Location
Chr.1: 210327873 - 210676298 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122834.3 NP_001116306.1 9 - 10 1221 137
NM_001170564.2 NP_001164035.1 7 - 8 810 137
NM_001170580.2 NP_001164051.1 9 - 10 1276 137
NM_001170587.2 NP_001164058.1 8 - 9 1153 137
NM_001170588.2 NP_001164059.1 8 - 9 1092 137
NM_018194.5 NP_060664.2 9 - 10 1287 137
XM_006711441.3 XP_006711504.1 10 - 11 1348 137
XM_011509737.2 XP_011508039.1 10 - 11 1348 137
XM_011509738.2 XP_011508040.1 8 - 9 1106 137
XM_011509739.2 XP_011508041.1 9 - 10 1273 137
XM_011509740.2 XP_011508042.1 10 - 11 1477 137
XM_011509741.2 XP_011508043.1 9 - 10 1221 137
XM_011509742.2 XP_011508044.1 9 - 10 1130 137
XM_011509744.2 XP_011508046.1 8 - 9 1281 137
XM_011509746.2 XP_011508048.1 8 - 9 1107 137
XM_011509747.1 XP_011508049.1 7 - 8 1071 137
XM_017001726.1 XP_016857215.1 8 - 9 1322 137
XM_017001727.1 XP_016857216.1 8 - 9 1322 137
XM_017001728.1 XP_016857217.1 9 - 10 1431 137
XM_017001729.1 XP_016857218.1 9 - 10 1152 137
XM_017001730.1 XP_016857219.1 8 - 9 1234 137
XM_017001731.1 XP_016857220.1 8 - 9 1040 137
XM_017001732.1 XP_016857221.1 8 - 9 917 137
XM_017001733.1 XP_016857222.1 7 - 8 1194 137
XM_017001734.1 XP_016857223.1 8 - 9 1228 137
XM_017001735.1 XP_016857224.1 8 - 9 1557 137
XM_017001736.1 XP_016857225.1 8 - 9 1128 137
XM_017001737.1 XP_016857226.1 7 - 8 701 137
XM_017001738.1 XP_016857227.1 7 - 8 808 137
XM_017001739.1 XP_016857228.1 7 - 8 941 137
XM_017001740.1 XP_016857229.1 7 - 8 1442 137
XM_017001741.1 XP_016857230.1 5 - 6 603 137
XM_017001742.1 XP_016857231.1 5 - 6 1242 137
XM_017001743.1 XP_016857232.1 5 - 6 599 137
XM_017001744.1 XP_016857233.1 5 - 6 1031 137
GenBank mRNA AK001586.1 - 9 - 10 1241 137
AK092186.1 - 3 - 4 360 137
AK295130.1 - 8 - 9 1173 137
AK297193.1 - 7 - 8 808 137
AK298991.1 - 8 - 9 1046 137
AK299200.1 - 7 - 8 1071 137
AK302955.1 - 8 - 9 1106 137
AK316190.1 - 8 - 9 1107 137
AK316524.1 - 8 - 9 1139 137
BC039071.1 - 9 - 10 1226 137
BC117130.1 - 8 - 9 1076 137

View Full Product Details