The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002800.2 | NP_001002800.1 | 14 - 15 | 2694 | 153 |
NM_001288753.1 | NP_001275682.1 | 14 - 15 | 2619 | 153 | |
NM_005496.3 | NP_005487.3 | 13 - 14 | 2511 | 153 | |
XM_006713459.3 | XP_006713522.1 | 13 - 14 | 2230 | 153 | |
XM_011512311.2 | XP_011510613.1 | 14 - 15 | 2582 | 153 | |
XM_011512312.2 | XP_011510614.1 | 13 - 14 | 2203 | 153 | |
XM_017005491.1 | XP_016860980.1 | 12 - 13 | 2425 | 153 | |
GenBank mRNA | AB019987.1 | - | 14 - 15 | 2355 | 153 |
AF092564.1 | - | 12 - 13 | 1985 | 153 | |
AK122939.1 | - | 6 - 7 | 153 | ||
AK128143.1 | - | 14 - 15 | 2273 | 153 | |
AK307508.1 | - | - | 1810 | 153 | |
AK315070.1 | - | 14 - 15 | 2350 | 153 | |
AL136877.1 | - | 13 - 14 | 2475 | 153 | |
AL833949.1 | - | 13 - 14 | 3402 | 153 |