The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321786.1 | NP_001308715.1 | 12 - 13 | 2185 | 105 |
NM_001321788.1 | NP_001308717.1 | 12 - 13 | 2104 | 105 | |
NM_001321789.1 | NP_001308718.1 | 12 - 13 | 2123 | 105 | |
NM_001321790.1 | NP_001308719.1 | 12 - 13 | 2058 | 105 | |
NM_001321791.1 | NP_001308720.1 | 12 - 13 | 2104 | 105 | |
NM_001321793.1 | NP_001308722.1 | 12 - 13 | 2280 | 105 | |
NM_001321794.1 | NP_001308723.1 | 12 - 13 | 2104 | 105 | |
NM_001321795.1 | NP_001308724.1 | 12 - 13 | 2280 | 105 | |
NM_001321796.1 | NP_001308725.1 | 11 - 12 | 1957 | 105 | |
NM_001321797.1 | NP_001308726.1 | 12 - 13 | 2104 | 105 | |
NM_001321798.1 | NP_001308727.1 | 12 - 13 | 2280 | 105 | |
NM_001321799.1 | NP_001308728.1 | 11 - 12 | 2133 | 105 | |
NM_001321806.1 | NP_001308735.1 | 11 - 12 | 1947 | 105 | |
NM_001321807.1 | NP_001308736.1 | 13 - 14 | 2424 | 105 | |
NM_001321808.1 | NP_001308737.1 | 13 - 14 | 2248 | 105 | |
NM_001321811.1 | NP_001308740.1 | 11 - 12 | 2123 | 105 | |
NM_001321813.1 | NP_001308742.1 | 13 - 14 | 2161 | 105 | |
NM_001321816.1 | NP_001308745.1 | 11 - 12 | 2123 | 105 | |
NM_001321820.1 | NP_001308749.1 | 14 - 15 | 2412 | 105 | |
NM_001321822.1 | NP_001308751.1 | 13 - 14 | 2330 | 105 | |
NM_170662.4 | NP_733762.2 | 12 - 13 | 2280 | 105 | |
NR_135806.1 | 13 - 14 | 2412 | 105 | ||
NR_135807.1 | 13 - 14 | 2328 | 105 | ||
NR_135808.1 | 12 - 13 | 2280 | 105 | ||
NR_135809.1 | 13 - 14 | 2394 | 105 | ||
NR_135810.1 | 12 - 13 | 2058 | 105 | ||
NR_135811.1 | 12 - 13 | 2104 | 105 | ||
NR_135812.1 | 12 - 13 | 2000 | 105 | ||
XM_011513257.1 | XP_011511559.1 | 12 - 13 | 2100 | 105 | |
XM_011513259.2 | XP_011511561.2 | 12 - 13 | 2225 | 105 | |
XM_017007395.1 | XP_016862884.1 | 12 - 13 | 2052 | 105 | |
XM_017007396.1 | XP_016862885.1 | 12 - 13 | 2223 | 105 | |
XM_017007397.1 | XP_016862886.1 | 11 - 12 | 2077 | 105 | |
XM_017007398.1 | XP_016862887.1 | 11 - 12 | 1903 | 105 | |
XM_017007399.1 | XP_016862888.1 | 11 - 12 | 2128 | 105 | |
XM_017007400.1 | XP_016862889.1 | 11 - 12 | 2077 | 105 | |
GenBank mRNA | AB075490.1 | - | 1 - 2 | 563 | 105 |
AK094184.1 | - | 3 - 4 | 506 | 105 | |
AK292792.1 | - | 12 - 13 | 2221 | 105 | |
AK302533.1 | - | 12 - 13 | 2058 | 105 | |
AK316276.1 | - | 12 - 13 | 2038 | 105 | |
BC032851.2 | - | 12 - 13 | 2221 | 105 | |
BX649066.1 | - | 3 - 4 | 328 | 105 | |
CR749655.1 | - | 1 - 2 | 107 | 105 | |
DQ349203.1 | - | 11 - 12 | 1958 | 105 | |
DQ349204.1 | - | 11 - 12 | 1958 | 105 | |
U26710.1 | - | 12 - 13 | 2280 | 105 | |
U26711.1 | - | 12 - 13 | 2280 | 105 | |
U26712.1 | - | 12 - 13 | 2280 | 105 |