Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FMNL2
Entrez Gene ID
114793
Gene Name
formin like 2
Gene Aliases
FHOD2
Chromosome Location
Chr.2: 152335170 - 152649834 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052905.3 NP_443137.2 17 - 18 2539 90
XM_005246263.2 XP_005246320.1 17 - 18 2606 90
XM_005246265.3 XP_005246322.1 17 - 18 2606 90
XM_011510530.1 XP_011508832.1 18 - 19 2627 90
XM_011510531.1 XP_011508833.1 18 - 19 2627 90
XM_011510532.2 XP_011508834.1 18 - 19 2627 90
XM_011510533.2 XP_011508835.1 18 - 19 2627 90
XM_011510534.1 XP_011508836.1 18 - 19 2627 90
XM_011510535.2 XP_011508837.1 17 - 18 2474 90
XM_011510536.2 XP_011508838.1 12 - 13 1578 90
GenBank mRNA AB067489.1 - 17 - 18 2310 90
AK094865.1 - 1 - 2 94 90
AK131316.1 - 3 - 4 737 90
AL834396.1 - 15 - 16 1923 90
BC113878.1 - 4 - 5 501 90
BC114438.1 - 4 - 5 506 90
BC167159.1 - 17 - 18 2310 90

View Full Product Details