Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FMNL2
Entrez Gene ID
114793
Gene Name
formin like 2
Gene Aliases
FHOD2
Chromosome Location
Chr.2: 152335170 - 152649834 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052905.3 NP_443137.2 21 - 22 3049 150
XM_005246263.2 XP_005246320.1 21 - 22 3116 150
XM_005246265.3 XP_005246322.1 21 - 22 3116 150
XM_011510530.1 XP_011508832.1 22 - 23 3137 150
XM_011510531.1 XP_011508833.1 22 - 23 3137 150
XM_011510532.2 XP_011508834.1 22 - 23 3137 150
XM_011510533.2 XP_011508835.1 22 - 23 3137 150
XM_011510534.1 XP_011508836.1 21 - 22 3068 150
XM_011510535.2 XP_011508837.1 21 - 22 2984 150
XM_011510536.2 XP_011508838.1 16 - 17 2088 150
GenBank mRNA AB067489.1 - 21 - 22 2820 150
AK094865.1 - 5 - 6 604 150
AK131316.1 - 7 - 8 1247 150
AL834396.1 - 19 - 20 2433 150
BC113878.1 - 8 - 9 1011 150
BC114438.1 - 8 - 9 1016 150
BC167159.1 - 21 - 22 2820 150

View Full Product Details