The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052905.3 | NP_443137.2 | 9 - 10 | 1239 | 81 |
XM_005246263.2 | XP_005246320.1 | 9 - 10 | 1306 | 81 | |
XM_005246265.3 | XP_005246322.1 | 9 - 10 | 1306 | 81 | |
XM_011510530.1 | XP_011508832.1 | 9 - 10 | 1306 | 81 | |
XM_011510531.1 | XP_011508833.1 | 9 - 10 | 1306 | 81 | |
XM_011510532.2 | XP_011508834.1 | 9 - 10 | 1306 | 81 | |
XM_011510533.2 | XP_011508835.1 | 9 - 10 | 1306 | 81 | |
XM_011510534.1 | XP_011508836.1 | 9 - 10 | 1306 | 81 | |
XM_011510535.2 | XP_011508837.1 | 8 - 9 | 1153 | 81 | |
XM_011510536.2 | XP_011508838.1 | 3 - 4 | 257 | 81 | |
GenBank mRNA | AB067489.1 | - | 9 - 10 | 1007 | 81 |
AL834396.1 | - | 7 - 8 | 620 | 81 | |
BC167159.1 | - | 9 - 10 | 1007 | 81 |