The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199173.1 | NP_001186102.1 | 2 - 3 | 497 | 75 |
NM_001199174.1 | NP_001186103.1 | 2 - 3 | 336 | 75 | |
NM_022372.4 | NP_071767.3 | 2 - 3 | 527 | 75 | |
XM_005255475.3 | XP_005255532.2 | 2 - 3 | 285 | 75 | |
XM_005255478.2 | XP_005255535.1 | 2 - 3 | 490 | 75 | |
XM_005255479.2 | XP_005255536.1 | 2 - 3 | 417 | 75 | |
XM_017023548.1 | XP_016879037.1 | 2 - 3 | 479 | 75 | |
XM_017023549.1 | XP_016879038.1 | 2 - 3 | 220 | 75 | |
XM_017023550.1 | XP_016879039.1 | 2 - 3 | 494 | 75 | |
XM_017023551.1 | XP_016879040.1 | 2 - 3 | 250 | 75 | |
XM_017023552.1 | XP_016879041.1 | 2 - 3 | 500 | 75 | |
GenBank mRNA | AK022227.1 | - | 2 - 3 | 306 | 75 |
AK098762.1 | - | 2 - 3 | 432 | 75 | |
AK302201.1 | - | 2 - 3 | 249 | 75 | |
AK304390.1 | - | 2 - 3 | 615 | 75 | |
AU121635.1 | - | 2 - 3 | 306 | 75 | |
AY223837.1 | - | 2 - 3 | 250 | 75 | |
BC001313.2 | - | 2 - 3 | 217 | 75 | |
BC017119.1 | - | 1 - 2 | 228 | 75 | |
BC052292.1 | - | 1 - 2 | 751 | 75 | |
BC088354.1 | - | 2 - 3 | 245 | 75 | |
BI761765.1 | - | 2 - 3 | 255 | 75 | |
DA382215.1 | - | 2 - 3 | 501 | 75 |