The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126105.2 | NP_001119577.1 | 6 - 7 | 1136 | 99 |
NM_001126106.2 | NP_001119578.1 | 6 - 7 | 1242 | 99 | |
NR_040448.1 | 5 - 6 | 1509 | 99 | ||
XM_006720302.1 | XP_006720365.1 | 6 - 7 | 1233 | 99 | |
XM_011537298.2 | XP_011535600.1 | 5 - 6 | 1144 | 99 | |
XM_011537299.1 | XP_011535601.1 | 5 - 6 | 1095 | 99 | |
GenBank mRNA | AB011263.1 | - | 4 - 5 | 894 | 99 |
AB020532.1 | - | 5 - 6 | 1420 | 99 | |
AF092032.1 | - | 6 - 7 | 1180 | 99 | |
AJ130718.1 | - | 6 - 7 | 1187 | 99 | |
AK025377.1 | - | 5 - 6 | 974 | 99 | |
AK301219.1 | - | 6 - 7 | 969 | 99 | |
AK309128.1 | - | 4 - 5 | 572 | 99 | |
AK314351.1 | - | 5 - 6 | 1050 | 99 | |
AL365451.1 | - | 4 - 5 | 552 | 99 | |
BC003062.2 | - | 5 - 6 | 1032 | 99 | |
BC010107.2 | - | 5 - 6 | 1026 | 99 | |
BX161519.1 | - | 6 - 7 | 1101 | 99 | |
BX248291.1 | - | 5 - 6 | 1030 | 99 | |
KU178460.1 | - | 4 - 5 | 894 | 99 | |
Y18474.2 | - | 6 - 7 | 1135 | 99 |