Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNL4B
Entrez Gene ID
54957
Gene Name
thioredoxin like 4B
Gene Aliases
DLP, Dim2
Chromosome Location
Chr.16: 72044261 - 72094316 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324354.1 NP_001311283.1 1 - 2 285 95
NM_001324355.1 NP_001311284.1 1 - 2 285 95
NM_017853.2 NP_060323.1 1 - 2 274 95
XM_017023377.1 XP_016878866.1 1 - 2 290 95
GenBank mRNA AK000518.1 - 1 - 2 249 95
AK222889.1 - 1 - 2 241 95
AK308891.1 - 1 - 2 191 95
BC009646.1 - 1 - 2 215 95
BM849401.1 - 1 - 2 186 95
BP355646.1 - 1 - 2 274 95

View Full Product Details