Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATAD1
Entrez Gene ID
84896
Gene Name
ATPase family, AAA domain containing 1
Gene Aliases
AFDC1, FNP001, THORASE
Chromosome Location
Chr.10: 87751512 - 87841345 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005270252.4 XP_005270309.1 1 - 2 157 70

View Full Product Details