Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LINC00588
- Entrez Gene ID
-
26138
- Gene Name
- long intergenic non-protein coding RNA 588
- Gene Aliases
-
C8orf71
- Chromosome Location
-
Chr.8: 57279543 - 57284731 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026772.1 |
|
3 - 4 |
1582 |
95 |
GenBank mRNA |
AL080200.1 |
- |
3 - 4 |
1582 |
95 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AW072317.1 |
- |
1 - 2 |
|
|
Chr. 8 - 57282194 - 57284731 [-] on Build GRCh38 |
95 |
GenBankmRNA |
AI672520.1 |
- |
1 - 2 |
|
|
Chr. 8 - 57282064 - 57284731 [-] on Build GRCh38 |
95 |
GenBankmRNA |
AW182665.1 |
- |
1 - 2 |
|
|
Chr. 8 - 57282130 - 57284731 [-] on Build GRCh38 |
95 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC286177 |
LOC286177 |
|
-17.6kb
|
Anti-sense |
Chr. - 57261226 - 57266611 on Build GRCh38 |
Downstream |
uncharacterized LOC101929488 |
LOC101929488 |
|
59.1kb
|
Anti-sense |
Chr. - 57343787 - 57364857 on Build GRCh38 |
View Full Product Details