Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALB
Entrez Gene ID
213
Gene Name
albumin
Gene Aliases
ANALBA, FDAH, PRO0883, PRO0903, PRO1341
Chromosome Location
Chr.4: 73404255 - 73421412 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000477.5 NP_000468.1 2 - 3 208 80
GenBank mRNA A06977.1 - 2 - 3 210 80
AF190168.1 - 2 - 3 135 80
AK292755.1 - 2 - 3 173 77
AK298437.1 - 2 - 3 201 80
AK298461.1 - 2 - 3 176 80
AK308044.1 - 2 - 3 176 80
AK314794.1 - 2 - 3 176 80
AY358313.1 - 2 - 3 158 80
AY550967.1 - 2 - 3 142 80
AY728024.1 - 2 - 3 141 80
AY960291.1 - 2 - 3 167 80
BC034023.1 - 2 - 3 176 80
BC035969.1 - 2 - 3 176 80
BC036003.1 - 2 - 3 176 80
BJ993972.1 - 2 - 3 118 80
DA536463.1 - 2 - 3 177 80
DA642320.1 - 2 - 3 208 80
DQ986150.1 - 2 - 3 146 80
HQ537426.1 - 1 - 2 63 80
V00494.1 - 2 - 3 174 80
V00495.1 - 2 - 3 210 80

View Full Product Details