Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NME5
Entrez Gene ID
8382
Gene Name
NME/NM23 family member 5
Gene Aliases
NM23-H5, NM23H5, RSPH23
Chromosome Location
Chr.5: 138115172 - 138139443 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003551.2 NP_003542.1 2 - 3 208 165
XM_005272099.1 XP_005272156.1 1 - 2 135 165
XM_006714716.3 XP_006714779.1 1 - 2 135 165
XM_017009945.1 XP_016865434.1 1 - 2 135 165
GenBank mRNA AF067724.1 - 2 - 3 178 165
BC026182.2 - 2 - 3 180 165
KU178416.1 - 1 - 2 130 165
U90450.1 - 2 - 3 154 165
Y14992.1 - 1 - 2 144 165

View Full Product Details