The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201628.2 | NP_963922.2 | 10 - 11 | 1831 | 82 |
XM_005245795.4 | XP_005245852.1 | 12 - 13 | 2086 | 82 | |
XM_011541074.2 | XP_011539376.1 | 12 - 13 | 2093 | 82 | |
XM_011541075.2 | XP_011539377.1 | 10 - 11 | 1749 | 82 | |
XM_011541076.2 | XP_011539378.1 | 10 - 11 | 1894 | 82 | |
XM_011541077.2 | XP_011539379.1 | 10 - 11 | 2873 | 82 | |
XM_011541081.2 | XP_011539383.1 | 4 - 5 | 573 | 82 | |
GenBank mRNA | AK293806.1 | - | 5 - 6 | 637 | 82 |
DN831635.1 | - | 1 - 2 | 113 | 82 |