The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199633.1 | NP_001186562.1 | 13 - 14 | 1591 | 78 |
NM_022127.2 | NP_071410.1 | 14 - 15 | 1537 | 78 | |
NR_037638.2 | 14 - 15 | 1767 | 78 | ||
XM_011518905.2 | XP_011517207.1 | 13 - 14 | 1661 | 78 | |
XM_011518906.2 | XP_011517208.1 | 14 - 15 | 1757 | 78 | |
XM_011518907.2 | XP_011517209.1 | 12 - 13 | 1476 | 78 | |
XM_011518908.2 | XP_011517210.1 | 8 - 9 | 1496 | 78 | |
GenBank mRNA | AF305210.1 | - | 14 - 15 | 1537 | 78 |
AK292849.1 | - | 13 - 14 | 1590 | 78 | |
AK304406.1 | - | 14 - 15 | 1747 | 78 | |
AK314039.1 | - | 13 - 14 | 1571 | 78 | |
BC093821.1 | - | 13 - 14 | 1474 | 78 | |
BC093823.1 | - | 13 - 14 | 1474 | 78 |