The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001251882.1 | NP_001238811.1 | 12 - 13 | 1638 | 96 |
NM_001251883.1 | NP_001238812.1 | 8 - 9 | 898 | 96 | |
NM_001251884.1 | NP_001238813.1 | 10 - 11 | 1152 | 96 | |
NM_001251885.1 | NP_001238814.1 | 11 - 12 | 1215 | 96 | |
NM_004624.3 | NP_004615.2 | 11 - 12 | 1261 | 96 | |
XM_005265437.2 | XP_005265494.1 | 11 - 12 | 1227 | 96 | |
XM_005265438.3 | XP_005265495.1 | 11 - 12 | 1790 | 96 | |
XM_005265439.2 | XP_005265496.1 | 10 - 11 | 1124 | 96 | |
XM_011534079.1 | XP_011532381.1 | 12 - 13 | 1989 | 96 | |
GenBank mRNA | AK056819.1 | - | 12 - 13 | 1638 | 96 |
AK123943.1 | - | 1 - 2 | 886 | 96 | |
AK126759.1 | - | 1 - 2 | 2236 | 96 | |
AK293548.1 | - | 8 - 9 | 893 | 96 | |
AK298115.1 | - | 11 - 12 | 1184 | 96 | |
AK314334.1 | - | 11 - 12 | 1230 | 96 | |
BC064424.1 | - | 11 - 12 | 1217 | 96 | |
EF577396.1 | - | 11 - 12 | 1137 | 96 | |
L13288.1 | - | 11 - 12 | 1231 | 96 | |
L20295.1 | - | 10 - 11 | 1041 | 96 | |
X75299.1 | - | 11 - 12 | 1157 | 96 |