The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015258.1 | NP_056073.1 | 21 - 22 | 2340 | 78 |
XM_006717018.2 | XP_006717081.1 | 21 - 22 | 2227 | 78 | |
XM_006717019.1 | XP_006717082.1 | 20 - 21 | 2188 | 78 | |
XM_017014552.1 | XP_016870041.1 | 20 - 21 | 2158 | 78 | |
GenBank mRNA | AB014574.1 | - | 21 - 22 | 2267 | 78 |
AK301208.1 | - | 8 - 9 | 1094 | 78 | |
AK303074.1 | - | 20 - 21 | 2188 | 78 | |
BC077732.1 | - | 5 - 6 | 553 | 78 |