Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FKBP15
Entrez Gene ID
23307
Gene Name
FK506 binding protein 15
Gene Aliases
FKBP133, KIAA0674, PPP1R76
Chromosome Location
Chr.9: 113165036 - 113221361 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015258.1 NP_056073.1 9 - 10 981 97
XM_006717018.2 XP_006717081.1 9 - 10 898 97
XM_006717019.1 XP_006717082.1 8 - 9 829 97
XM_017014552.1 XP_016870041.1 8 - 9 829 97
GenBank mRNA AB014574.1 - 9 - 10 908 97
AK303074.1 - 8 - 9 829 97
AK307498.1 - 8 - 9 810 97
AK307507.1 - 13 - 14 1415 97
BC009609.1 - 9 - 10 938 97

View Full Product Details