Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM2
Entrez Gene ID
23670
Gene Name
transmembrane protein 2
Gene Aliases
-
Chromosome Location
Chr.9: 71683366 - 71769532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135820.1 NP_001129292.1 12 - 13 2745 83
NM_013390.2 NP_037522.1 13 - 14 2934 83
XM_005251869.4 XP_005251926.1 13 - 14 2567 83
GenBank mRNA AB037833.2 - 13 - 14 2527 83
AF137030.1 - 13 - 14 2542 83
AK075370.1 - 6 - 7 662 83
BC140773.1 - 12 - 13 2247 83
BC144510.1 - 13 - 14 2402 83
BC146780.1 - 13 - 14 2527 83
BC171782.1 - 12 - 13 2248 83

View Full Product Details