Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM2
Entrez Gene ID
23670
Gene Name
transmembrane protein 2
Gene Aliases
-
Chromosome Location
Chr.9: 71683366 - 71769532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135820.1 NP_001129292.1 16 - 17 3331 76
NM_013390.2 NP_037522.1 17 - 18 3520 76
XM_005251869.4 XP_005251926.1 17 - 18 3153 76
GenBank mRNA AB037833.2 - 17 - 18 3113 76
AF137030.1 - 17 - 18 3128 76
AK075370.1 - 10 - 11 1248 76
BC140773.1 - 16 - 17 2833 76
BC144510.1 - 17 - 18 2988 76
BC146780.1 - 17 - 18 3113 76
BC171782.1 - 16 - 17 2834 76

View Full Product Details