The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170416.1 | NP_001163887.1 | 20 - 21 | 3090 | 153 |
NM_004817.3 | NP_004808.2 | 20 - 21 | 3315 | 153 | |
XM_005252314.2 | XP_005252371.1 | 20 - 21 | 3368 | 153 | |
XM_011519206.2 | XP_011517508.1 | 22 - 23 | 3660 | 153 | |
XM_011519207.2 | XP_011517509.1 | 21 - 22 | 3156 | 153 | |
XM_011519208.2 | XP_011517510.1 | 21 - 22 | 3116 | 153 | |
XM_011519209.2 | XP_011517511.1 | 19 - 20 | 2945 | 153 | |
XM_017015326.1 | XP_016870815.1 | 21 - 22 | 3213 | 153 | |
GenBank mRNA | AK295034.1 | - | 20 - 21 | 3090 | 153 |
AK304447.1 | - | 22 - 23 | 3065 | 153 | |
BC027592.1 | - | 20 - 21 | 3206 | 153 | |
L27476.1 | - | 20 - 21 | 153 |