The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170415.1 | NP_001163886.1 | 20 - 21 | 3379 | 78 |
NM_001170416.1 | NP_001163887.1 | 21 - 22 | 3420 | 78 | |
NM_004817.3 | NP_004808.2 | 21 - 22 | 3645 | 78 | |
XM_005252314.2 | XP_005252371.1 | 21 - 22 | 3698 | 78 | |
XM_011519204.1 | XP_011517506.1 | 20 - 21 | 3277 | 78 | |
XM_011519206.2 | XP_011517508.1 | 23 - 24 | 3990 | 78 | |
XM_011519207.2 | XP_011517509.1 | 22 - 23 | 3486 | 78 | |
XM_011519208.2 | XP_011517510.1 | 22 - 23 | 3446 | 78 | |
XM_011519209.2 | XP_011517511.1 | 20 - 21 | 3275 | 78 | |
XM_017015326.1 | XP_016870815.1 | 22 - 23 | 3543 | 78 | |
XM_017015327.1 | XP_016870816.1 | 20 - 21 | 3534 | 78 | |
GenBank mRNA | AK295034.1 | - | 21 - 22 | 3420 | 78 |
AK304447.1 | - | 23 - 24 | 3395 | 78 | |
AK307697.1 | - | 4 - 5 | 866 | 78 | |
BC027592.1 | - | 21 - 22 | 3536 | 78 | |
L27476.1 | - | 21 - 22 | 3410 | 78 | |
U84581.1 | - | 1 - 2 | 191 | 78 |