The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022897.4 | NP_075048.1 | 16 - 17 | 2013 | 126 |
XM_011534627.1 | XP_011532929.1 | 16 - 17 | 1869 | 126 | |
XM_011534631.2 | XP_011532933.1 | 15 - 16 | 1882 | 126 | |
XM_017009736.1 | XP_016865225.1 | 15 - 16 | 1749 | 126 | |
XM_017009737.1 | XP_016865226.1 | 15 - 16 | 1882 | 126 | |
XM_017009738.1 | XP_016865227.1 | 15 - 16 | 1722 | 126 | |
XM_017009739.1 | XP_016865228.1 | 15 - 16 | 1722 | 126 | |
XM_017009740.1 | XP_016865229.1 | 14 - 15 | 1602 | 126 | |
XM_017009741.1 | XP_016865230.1 | 16 - 17 | 1869 | 126 | |
XM_017009742.1 | XP_016865231.1 | 16 - 17 | 1869 | 126 | |
XM_017009743.1 | XP_016865232.1 | 16 - 17 | 2043 | 126 | |
XM_017009744.1 | XP_016865233.1 | 16 - 17 | 1869 | 126 | |
XM_017009745.1 | XP_016865234.1 | 8 - 9 | 1066 | 126 | |
XM_017009747.1 | XP_016865236.1 | 4 - 5 | 341 | 126 | |
GenBank mRNA | AF222747.1 | - | 16 - 17 | 1869 | 126 |
AJ271459.1 | - | 16 - 17 | 1877 | 126 | |
AJ288952.1 | - | 19 - 20 | 2216 | 126 | |
AJ288953.1 | - | 18 - 19 | 2055 | 126 | |
AJ288954.1 | - | 18 - 19 | 2176 | 126 | |
AJ288955.1 | - | 17 - 18 | 1995 | 126 | |
AK027880.1 | - | 14 - 15 | 1697 | 126 | |
AK315159.1 | - | 17 - 18 | 2081 | 126 | |
AL832980.1 | - | 16 - 17 | 1912 | 126 |