Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RANBP17
Entrez Gene ID
64901
Gene Name
RAN binding protein 17
Gene Aliases
-
Chromosome Location
Chr.5: 170861882 - 171300015 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022897.4 NP_075048.1 7 - 8 910 104
XM_011534627.1 XP_011532929.1 7 - 8 766 104
XM_011534631.2 XP_011532933.1 6 - 7 779 104
XM_011534636.2 XP_011532938.1 7 - 8 766 104
XM_011534637.2 XP_011532939.1 7 - 8 766 104
XM_017009736.1 XP_016865225.1 7 - 8 766 104
XM_017009737.1 XP_016865226.1 6 - 7 779 104
XM_017009738.1 XP_016865227.1 7 - 8 766 104
XM_017009739.1 XP_016865228.1 7 - 8 766 104
XM_017009740.1 XP_016865229.1 7 - 8 766 104
XM_017009741.1 XP_016865230.1 7 - 8 766 104
XM_017009742.1 XP_016865231.1 7 - 8 766 104
XM_017009743.1 XP_016865232.1 7 - 8 940 104
XM_017009744.1 XP_016865233.1 7 - 8 766 104
XM_017009746.1 XP_016865235.1 7 - 8 766 104
GenBank mRNA AF222747.1 - 7 - 8 766 104
AJ271459.1 - 7 - 8 774 104
AJ288952.1 - 7 - 8 774 104
AJ288953.1 - 7 - 8 774 104
AJ288954.1 - 7 - 8 774 104
AJ288955.1 - 7 - 8 774 104
AK027880.1 - 5 - 6 594 104
AK315159.1 - 7 - 8 860 104
AL832980.1 - 6 - 7 691 104

View Full Product Details