The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025153.2 | NP_079429.2 | 23 - 24 | 4492 | 106 |
XM_006714833.2 | XP_006714896.1 | 22 - 23 | 4073 | 106 | |
XM_011534468.2 | XP_011532770.1 | 22 - 23 | 4049 | 106 | |
XM_011534469.1 | XP_011532771.1 | 23 - 24 | 4313 | 106 | |
XM_011534472.2 | XP_011532774.1 | 18 - 19 | 3789 | 106 | |
XM_017009252.1 | XP_016864741.1 | 22 - 23 | 4013 | 106 | |
XM_017009253.1 | XP_016864742.1 | 21 - 22 | 3872 | 106 | |
GenBank mRNA | AB018258.2 | - | 23 - 24 | 4319 | 106 |
AK025290.1 | - | 8 - 9 | 1415 | 106 | |
AK025294.1 | - | 5 - 6 | 731 | 106 |