Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP10B
Entrez Gene ID
23120
Gene Name
ATPase phospholipid transporting 10B (putative)
Gene Aliases
ATPVB
Chromosome Location
Chr.5: 160563120 - 160929411 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025153.2 NP_079429.2 14 - 15 2659 68
XM_006714833.2 XP_006714896.1 13 - 14 2240 68
XM_011534468.2 XP_011532770.1 13 - 14 2216 68
XM_011534469.1 XP_011532771.1 14 - 15 2480 68
XM_011534472.2 XP_011532774.1 9 - 10 1956 68
XM_017009252.1 XP_016864741.1 13 - 14 2180 68
XM_017009253.1 XP_016864742.1 13 - 14 2240 68
GenBank mRNA AB018258.2 - 14 - 15 2486 68
BC108322.1 - 3 - 4 636 68

View Full Product Details