The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252156.1 | NP_001239085.1 | 14 - 15 | 2075 | 114 |
NM_015465.4 | NP_056280.2 | 14 - 15 | 2078 | 114 | |
GenBank mRNA | AK022748.1 | - | 14 - 15 | 2025 | 114 |
AK074066.2 | - | 14 - 15 | 2050 | 114 | |
AK315101.1 | - | 14 - 15 | 2063 | 114 | |
AL117665.1 | - | 9 - 10 | 1140 | 114 | |
AY063750.1 | - | 14 - 15 | 1995 | 114 | |
BC008776.2 | - | 2 - 3 | 236 | 114 | |
BC014147.2 | - | 2 - 3 | 236 | 114 | |
BC036894.1 | - | 14 - 15 | 2058 | 114 | |
BC040931.1 | - | 14 - 15 | 2048 | 114 | |
BC113614.1 | - | 14 - 15 | 2034 | 114 | |
BC143724.1 | - | 14 - 15 | 2031 | 114 |