The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197294.1 | NP_001184223.1 | 5 - 6 | 1084 | 153 |
NM_001387.2 | NP_001378.1 | 5 - 6 | 654 | 153 | |
XM_011537574.1 | XP_011535876.1 | 6 - 7 | 1038 | 153 | |
GenBank mRNA | BC039006.1 | - | 5 - 6 | 620 | 153 |
BC077077.1 | - | 5 - 6 | 1084 | 153 | |
D78014.1 | - | 5 - 6 | 654 | 153 | |
EU007906.1 | - | 5 - 6 | 886 | 153 | |
Y07818.1 | - | 5 - 6 | 675 | 153 |