Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIF13A
Entrez Gene ID
63971
Gene Name
kinesin family member 13A
Gene Aliases
RBKIN, bA500C11.2
Chromosome Location
Chr.6: 17759183 - 17987623 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105566.2 NP_001099036.1 32 - 33 4104 129
NM_001105567.2 NP_001099037.1 31 - 32 4065 129
NM_001105568.2 NP_001099038.1 31 - 32 4065 129
NM_022113.5 NP_071396.4 32 - 33 4104 129
XM_017011188.1 XP_016866677.1 32 - 33 4099 129
XM_017011189.1 XP_016866678.1 31 - 32 4060 129
XM_017011190.1 XP_016866679.1 32 - 33 4099 129
XM_017011191.1 XP_016866680.1 30 - 31 4131 129
XM_017011192.1 XP_016866681.1 32 - 33 4099 129
XM_017011193.1 XP_016866682.1 29 - 30 4007 129
GenBank mRNA AJ291578.1 - 31 - 32 3905 129
AJ291579.1 - 31 - 32 3905 129
AK025303.1 - 16 - 17 2125 129
AK225214.1 - 16 - 17 2125 129
AY014403.1 - 32 - 33 3963 129
AY014404.1 - 32 - 33 3963 129
BC127115.1 - 32 - 33 3962 129
BC127116.1 - 31 - 32 3923 129

View Full Product Details