The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003883.3 | NP_003874.2 | 2 - 3 | 209 | 80 |
GenBank mRNA | AF039703.1 | - | 2 - 3 | 165 | 80 |
BC000614.1 | - | 2 - 3 | 198 | 80 | |
BM461585.1 | - | 2 - 3 | 199 | 80 | |
U66914.1 | - | 2 - 3 | 154 | 80 | |
U75696.1 | - | - | 57 | 80 | |
U75697.1 | - | 2 - 3 | 198 | 80 |