The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164326.1 | NP_001157798.1 | 17 - 18 | 2318 | 83 |
NM_001300961.1 | NP_001287890.1 | 17 - 18 | 2314 | 83 | |
NM_001300962.1 | NP_001287891.1 | 16 - 17 | 2239 | 83 | |
NM_001300966.1 | NP_001287895.1 | 18 - 19 | 2406 | 83 | |
NM_006696.3 | NP_006687.3 | 18 - 19 | 2528 | 83 | |
NM_139199.1 | NP_631938.1 | 17 - 18 | 2309 | 83 | |
XM_005271855.2 | XP_005271912.1 | 18 - 19 | 2519 | 83 | |
XM_005271856.2 | XP_005271913.1 | 18 - 19 | 2519 | 83 | |
XM_005271857.2 | XP_005271914.1 | 17 - 18 | 2309 | 83 | |
XM_005271859.3 | XP_005271916.1 | 16 - 17 | 2090 | 83 | |
XM_005271860.3 | XP_005271917.1 | 18 - 19 | 2519 | 83 | |
XM_005271861.3 | XP_005271918.1 | 17 - 18 | 2300 | 83 | |
XM_011543110.1 | XP_011541412.1 | 18 - 19 | 2505 | 83 | |
XM_017008972.1 | XP_016864461.1 | 17 - 18 | 2309 | 83 | |
XM_017008973.1 | XP_016864462.1 | 17 - 18 | 2300 | 83 | |
XM_017008976.1 | XP_016864465.1 | 16 - 17 | 2090 | 83 | |
GenBank mRNA | AB209079.1 | - | 18 - 19 | 2474 | 83 |
AF016270.1 | - | 17 - 18 | 2460 | 83 | |
AK289951.1 | - | 18 - 19 | 2488 | 83 | |
AK293621.1 | - | 16 - 17 | 2199 | 83 | |
AK297608.1 | - | 8 - 9 | 1597 | 83 | |
AK297756.1 | - | 17 - 18 | 2274 | 83 | |
AK312333.1 | - | 18 - 19 | 2488 | 83 | |
AL136823.1 | - | 17 - 18 | 2282 | 83 | |
BC008039.2 | - | 18 - 19 | 2470 | 83 | |
BC008076.2 | - | 18 - 19 | 2470 | 83 | |
X87613.1 | - | 17 - 18 | 2336 | 83 | |
X94234.1 | - | 16 - 17 | 2248 | 83 |