The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139199.1 | NP_631938.1 | 19 - 20 | 2590 | 82 |
XM_005271855.2 | XP_005271912.1 | 20 - 21 | 2800 | 82 | |
XM_005271857.2 | XP_005271914.1 | 19 - 20 | 2590 | 82 | |
XM_005271859.3 | XP_005271916.1 | 18 - 19 | 2371 | 82 | |
XM_011543109.1 | XP_011541411.1 | 18 - 19 | 2638 | 82 | |
XM_011543110.1 | XP_011541412.1 | 20 - 21 | 2786 | 82 | |
XM_017008969.1 | XP_016864458.1 | 17 - 18 | 2419 | 82 | |
GenBank mRNA | AL136823.1 | - | 19 - 20 | 2563 | 82 |