The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139199.1 | NP_631938.1 | 21 - 22 | 2908 | 67 |
XM_005271855.2 | XP_005271912.1 | 22 - 23 | 3118 | 67 | |
XM_005271856.2 | XP_005271913.1 | 21 - 22 | 3073 | 67 | |
XM_005271857.2 | XP_005271914.1 | 21 - 22 | 2908 | 67 | |
XM_005271859.3 | XP_005271916.1 | 20 - 21 | 2689 | 67 | |
XM_011543109.1 | XP_011541411.1 | 20 - 21 | 2956 | 67 | |
XM_011543110.1 | XP_011541412.1 | 22 - 23 | 3104 | 67 | |
XM_017008969.1 | XP_016864458.1 | 19 - 20 | 2737 | 67 | |
GenBank mRNA | AL136823.1 | - | 21 - 22 | 2881 | 67 |