The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139199.1 | NP_631938.1 | 25 - 26 | 3495 | 80 |
XM_005271855.2 | XP_005271912.1 | 26 - 27 | 3705 | 80 | |
XM_005271856.2 | XP_005271913.1 | 25 - 26 | 3660 | 80 | |
XM_005271857.2 | XP_005271914.1 | 25 - 26 | 3495 | 80 | |
XM_005271859.3 | XP_005271916.1 | 24 - 25 | 3276 | 80 | |
XM_011543109.1 | XP_011541411.1 | 24 - 25 | 3543 | 80 | |
XM_011543110.1 | XP_011541412.1 | 26 - 27 | 3691 | 80 | |
XM_017008969.1 | XP_016864458.1 | 23 - 24 | 3324 | 80 | |
GenBank mRNA | AL136823.1 | - | 25 - 26 | 3468 | 80 |