The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300961.1 | NP_001287890.1 | 8 - 9 | 704 | 70 |
NM_001300962.1 | NP_001287891.1 | 7 - 8 | 629 | 70 | |
NM_139199.1 | NP_631938.1 | 8 - 9 | 699 | 70 | |
XM_005271859.3 | XP_005271916.1 | 8 - 9 | 690 | 70 | |
XM_005271861.3 | XP_005271918.1 | 8 - 9 | 690 | 70 | |
XM_017008969.1 | XP_016864458.1 | 8 - 9 | 690 | 70 | |
XM_017008973.1 | XP_016864462.1 | 8 - 9 | 690 | 70 | |
XM_017008975.1 | XP_016864464.1 | 8 - 9 | 690 | 70 | |
XM_017008976.1 | XP_016864465.1 | 8 - 9 | 690 | 70 | |
GenBank mRNA | AK293621.1 | - | 7 - 8 | 589 | 70 |
AK297756.1 | - | 8 - 9 | 664 | 70 | |
AK310380.1 | - | 6 - 7 | 538 | 70 | |
AL136823.1 | - | 8 - 9 | 672 | 70 |