Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRAMD1A
Entrez Gene ID
57655
Gene Name
GRAM domain containing 1A
Gene Aliases
KIAA1533
Chromosome Location
Chr.19: 34994784 - 35026471 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020895.4 NP_065946.2 16 - 17 2004 61
XM_011527149.1 XP_011525451.1 16 - 17 2001 61
XM_011527153.1 XP_011525455.1 17 - 18 2177 61
XM_011527154.2 XP_011525456.1 16 - 17 1977 61
XM_011527155.1 XP_011525457.1 17 - 18 2255 61
XM_011527156.1 XP_011525458.1 15 - 16 1891 61
XM_017027034.1 XP_016882523.1 16 - 17 2004 61
GenBank mRNA BC014077.2 - 16 - 17 1992 61

View Full Product Details