The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144650.2 | NP_653251.2 | 6 - 7 | 584 | 69 |
GenBank mRNA | AK056992.1 | - | 5 - 6 | 521 | 69 |
AK293530.1 | - | 3 - 4 | 424 | 69 | |
AK300243.1 | - | 6 - 7 | 563 | 69 | |
AY033237.1 | - | 6 - 7 | 569 | 69 |