The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042539.2 | NP_001036004.1 | 4 - 5 | 1444 | 67 |
XM_006721047.3 | XP_006721110.1 | 5 - 6 | 1231 | 67 | |
GenBank mRNA | AK074720.1 | - | 1 - 2 | 70 | 67 |
BC041629.1 | - | 3 - 4 | 594 | 67 |