The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256269.1 | NP_001243198.1 | 11 - 12 | 1712 | 65 |
NM_001256270.1 | NP_001243199.1 | 10 - 11 | 1800 | 65 | |
NM_007317.2 | NP_015556.1 | 10 - 11 | 1663 | 65 | |
GenBank mRNA | AB017430.2 | - | 10 - 11 | 1640 | 65 |
AK223431.1 | - | 10 - 11 | 1649 | 65 | |
AK294380.1 | - | 10 - 11 | 2039 | 65 | |
AK312234.1 | - | 10 - 11 | 1654 | 65 | |
AK316050.1 | - | 10 - 11 | 1800 | 65 | |
AK316389.1 | - | 11 - 12 | 1671 | 65 | |
BC004352.1 | - | 10 - 11 | 1635 | 65 | |
BC028155.1 | - | 10 - 11 | 1621 | 65 | |
BT007259.1 | - | 10 - 11 | 1616 | 65 | |
L29096.1 | - | 1 - 2 | 133 | 65 |