Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIF22
Entrez Gene ID
3835
Gene Name
kinesin family member 22
Gene Aliases
A-328A3.2, KID, KNSL4, OBP, OBP-1, OBP-2, SEMDJL2
Chromosome Location
Chr.16: 29790062 - 29805385 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256269.1 NP_001243198.1 14 - 15 2047 70
NM_001256270.1 NP_001243199.1 13 - 14 2135 70
NM_007317.2 NP_015556.1 13 - 14 1998 70
GenBank mRNA AB017430.2 - 13 - 14 1975 70
AK223431.1 - 13 - 14 1984 70
AK294380.1 - 13 - 14 2374 70
AK312234.1 - 13 - 14 1989 70
AK316050.1 - 13 - 14 2135 70
AK316389.1 - 14 - 15 2006 70
BC004352.1 - 13 - 14 1970 70
BC028155.1 - 13 - 14 1956 70
BT007259.1 - 13 - 14 1951 70
L29095.1 - 2 - 3 288 70
L29096.1 - 4 - 5 468 70

View Full Product Details