Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MVP
Entrez Gene ID
9961
Gene Name
major vault protein
Gene Aliases
LRP, VAULT1
Chromosome Location
Chr.16: 29820394 - 29848039 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293205.1 NP_001280134.1 11 - 12 2101 96
NM_005115.4 NP_005106.2 13 - 14 2416 96
NM_017458.3 NP_059447.2 13 - 14 2457 96
GenBank mRNA AK293302.1 - 13 - 14 2311 96
AK298238.1 - 11 - 12 2075 96
AK302052.1 - 13 - 14 2163 96
BC015623.2 - 13 - 14 2324 96
BM999583.1 - 2 - 3 96
KJ534884.1 - 12 - 13 2264 96
KJ534885.1 - 11 - 12 2066 96
KJ535063.1 - 7 - 8 1350 96
X79882.2 - 13 - 14 2369 96

View Full Product Details